5-(Benzyloxy)-3-bromo-1H-indole, N-BOC protected 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 5-(benzyloxy)-3-bromo-1H-indole-1-carboxylate, 5-(Benzyloxy)-3-bromo-1-(tert-butoxycarbonyl)-1H-indole

OR1721-500MG
APOSOR1721-500MGEA 30.6 EUR
APOSOR1721-500MG
5-(Benzyloxy)-3-bromo-1H-indole, N-BOC protected 95%
5-(Benzyloxy)-3-bromo-1H-indole, N-BOC protected
Formel: C₂₀H₂₀BrNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05864770
CAS-Nummer: 914349-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR