5-(Benzyloxy)-3-iodo-1H-indole, N-BOC protected 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Benzyloxy)-1-(tert-butoxycarbonyl)-3-iodo-1H-indole, tert-Butyl 5-(benzyloxy)-3-iodo-1H-indole-1-carboxylate

OR1722-250MG
APOSOR1722-250MGEA 77 EUR
APOSOR1722-250MG
5-(Benzyloxy)-3-iodo-1H-indole, N-BOC protected 95+%
5-(Benzyloxy)-3-iodo-1H-indole, N-BOC protected
Formel: C₂₀H₂₀INO₃
Molecular Weight: 449,282 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864771
CAS-Nummer: 914349-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR