6-Benzyloxy-3-iodo-1H-indole, N-BOC protected 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Benzyloxy-3-iodo-1H-indole-1-carboxylic acid tert-butyl ester

OR1724-1G
APOSOR1724-1GEA 255 EUR
APOSOR1724-1G
6-Benzyloxy-3-iodo-1H-indole, N-BOC protected 95%
6-Benzyloxy-3-iodo-1H-indole, N-BOC protected
Formel: C₂₀H₂₀INO₃
Molecular Weight: 449,282 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864773
CAS-Nummer: 914349-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR