Methyl 4-bromothiophene-2-carboxylate 98%

Lieferant: Apollo Scientific

OR17259-10G
APOSOR17259-10GEA 42.2 EUR
APOSOR17259-10G
Methyl 4-bromothiophene-2-carboxylate 98%
Methyl 4-bromothiophene-2-carboxylate
Formel: C₆H₅BrO₂S
Molecular Weight: 221,072 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06203668
CAS-Nummer: 62224-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR