3-Bromo-5-nitro-1H-indole, N-BOC protected 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-bromo-5-nitro-1H-indole-1-carboxylate, 3-Bromo-1-(tert-butoxycarbonyl)-5-nitro-1H-indole

OR1727-250MG
APOSOR1727-250MGEA 34.8 EUR
APOSOR1727-250MG
3-Bromo-5-nitro-1H-indole, N-BOC protected 98%
3-Bromo-5-nitro-1H-indole, N-BOC protected
Formel: C₁₃H₁₃BrN₂O₄
Molecular Weight: 341,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864776
CAS-Nummer: 914349-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR