4-Chloro-1H-pyrazolo[3,4-d]pyrimidin-6-amine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 6-Amino-4-chloro-1H-pyrazolo[3,4-d]pyrimidine

OR17302-5G
APOSOR17302-5GEA 143 EUR
APOSOR17302-5G
4-Chloro-1H-pyrazolo[3,4-d]pyrimidin-6-amine
4-Chloro-1H-pyrazolo[3,4-d]pyrimidin-6-amine
Formel: C₅H₄ClN₅
Molecular Weight: 169.5700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08059805
CAS-Nummer: 100644-65-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR