5-Chlor-2-nitrophenol 98%

Lieferant: Apollo Scientific
Achtung

OR17307-1G
APOSOR17307-1GEA 52.2 EUR
APOSOR17307-1G
5-Chlor-2-nitrophenol 98%
5-Chlor-2-nitrophenol
Formel: C₆H₄ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09260849
CAS-Nummer: 611-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR