3-Bromo-2-methyl-1H-indole, N-BOC protected 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-1-(tert-butoxycarbonyl)-2-methyl-1H-indole, tert-Butyl 3-bromo-2-methyl-1H-indole-1-carboxylate, 1-Boc-3-Bromo-2-methylindole
OR1732-250MG
APOSOR1732-250MGEA
21.9
EUR
APOSOR1732-250MG
3-Bromo-2-methyl-1H-indole, N-BOC protected 98%
3-Bromo-2-methyl-1H-indole, N-BOC protected
Formel:
C₁₄H₁₆BrNO₂ Molecular Weight: 310,186 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05864780 CAS-Nummer: 775305-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...