1-Boc-3-iodoindole 98%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-iodo-1H-indole-1-carboxylate

OR1733-1G
APOSOR1733-1GEA 0 EUR
APOSOR1733-1G
1-Boc-3-iodoindole 98%
1-Boc-3-iodoindole
Formel: C₁₃H₁₄INO₂
Molecular Weight: 343,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864781
CAS-Nummer: 192189-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR