3-Iodo-2-methyl-1H-indole, N-BOC protected 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-iodo-2-methyl-1H-indole-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-iodo-2-methyl-1H-indole

OR1740-1G
APOSOR1740-1GEA 180 EUR
APOSOR1740-1G
3-Iodo-2-methyl-1H-indole, N-BOC protected 95%
3-Iodo-2-methyl-1H-indole, N-BOC protected
Formel: C₁₄H₁₆INO₂
Molecular Weight: 357.187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864787
CAS-Nummer: 877996-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR