3-(Methylsulfonyl)anilin

Lieferant: Apollo Scientific
Achtung

OR17400-1G OR17400-5G OR17400-10G
APOSOR17400-1GEA 34.8 EUR
APOSOR17400-1G APOSOR17400-5G APOSOR17400-10G
3-(Methylsulfonyl)anilin
3-(Methylsulfonyl)anilin
Formel: C₇H₉NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD03764428
CAS-Nummer: 35216-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR