2,5-Dichloro-4-methoxyaniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-2,5-dichloroanisole, 2,5-Dichloro-p-anisidine

OR17402-10G
APOSOR17402-10GEA 36.4 EUR
APOSOR17402-10G
2,5-Dichloro-4-methoxyaniline
2,5-Dichloro-4-methoxyaniline
Formel: C₇H₇Cl₂NO
Molecular Weight: 192,043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16606083
CAS-Nummer: 101251-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR