1-{[Isocyano(phenyl)methyl]sulfonyl}-4-methylbenzol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-Phenyl-1-tosyl)methyl Isocyanide, α-Tosylbenzyl Isocyanide, α-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-Tosylbenzyl Isocyanide

OR1744-1G OR1744-5G
APOSOR1744-1GEA 78.3 EUR
APOSOR1744-1G APOSOR1744-5G
1-{[Isocyano(phenyl)methyl]sulfonyl}-4-methylbenzol 97%
a-Tosylbenzyl isocyanide
Formel: C₁₅H₁₃NO₂S
Molecular Weight: 271.3342 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114776
CAS-Nummer: 36635-66-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR