6-Bromo-3-(dimethylamino)-1,2,3,4-tetrahydro-9H-carbazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-N,N-dimethyl-2,3,4,9-tetrahydro-1H-carbazol-3-amine

OR17505-1G
APOSOR17505-1GEA 232 EUR
APOSOR17505-1G
6-Bromo-3-(dimethylamino)-1,2,3,4-tetrahydro-9H-carbazole
6-Bromo-3-(dimethylamino)-1,2,3,4-tetrahydro-9H-carbazole

Useful as 5-HT1F receptor antagonists

Formel: C₁₄H₁₇BrN₂
Molecular Weight: 293,202 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13563140
CAS-Nummer: 186545-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR