(1-Acetylpiperidin-4-yl)acetic acid

Lieferant: Apollo Scientific

OR17507-1G OR17507-5G
APOSOR17507-1GEA 0 EUR
APOSOR17507-1G APOSOR17507-5G
(1-Acetylpiperidin-4-yl)acetic acid
(1-Acetylpiperidin-4-yl)acetic acid
Formel: C₉H₁₅NO₃
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12027293
CAS-Nummer: 885274-84-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR