2-Methyloxazole-4-carboxaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methyloxazole-4-carbaldehyde
OR17528-1G
APOSOR17528-1GEA
55.1
EUR
APOSOR17528-1G
2-Methyloxazole-4-carboxaldehyde 97%
2-Methyloxazole-4-carboxaldehyde
Formel:
C₅H₅NO₂ Molecular Weight: 111.1 g/mol Schmelzpunkt: 70…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03265461 CAS-Nummer: 113732-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...