2,6-Dichlor-3-methylpyridin 98%

Lieferant: Apollo Scientific
Achtung

OR17532-25G
APOSOR17532-25GEA 81.2 EUR
APOSOR17532-25G
2,6-Dichlor-3-methylpyridin 98%
2,6-Dichloro-3-methylpyridine
Formel: C₆H₅Cl₂N
Molecular Weight: 162.02 g/mol
Schmelzpunkt: 54…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00137847
CAS-Nummer: 58584-94-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR