2-Methyl-1,3-oxazole-4-carbohydrazide

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Hydrazinocarbonyl)-2-methyl-1,3-oxazole, 2-Methyl-1,3-oxazole-4-carboxylic acid hydrazide

OR17536-250MG
APOSOR17536-250MGEA 58 EUR
APOSOR17536-250MG
2-Methyl-1,3-oxazole-4-carbohydrazide
2-Methyl-1,3-oxazole-4-carbohydrazide
Formel: C₅H₇N₃O₂
Molecular Weight: 141,128 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11052651
CAS-Nummer: 500341-65-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR