2-Aminobenzothiazol-6-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Hydroxybenzothiazol-2-amine, 2-Amino-6-benzothiazolol, 2-Aminobenzothiazol-6-ol, 6-Hydroxy-2-aminobenzothiazole, 2-Amino-6-hydroxy-1,3-benzothiazole, 2-Amino-6-hydroxybenzothiazole

OR17549-1G
APOSOR17549-1GEA 58 EUR
APOSOR17549-1G
2-Aminobenzothiazol-6-ol
2-Aminobenzothiazol-6-ol
Formel: C₇H₆N₂OS
Molecular Weight: 166,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00511763
CAS-Nummer: 26278-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR