tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-BOC-4-(2-aminoethyl)piperazine, 4-(2-Aminoethyl)-1-boc-piperazine, tert-Butyl 4-(2-aminoethyl)piperazine-1-carboxylate

OR1755-25G
APOSOR1755-25GEA 790 EUR
APOSOR1755-25G
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate
Formel: C₁₁H₂₃N₃O₂
Lagertemperatur: Kühlschrank
MDL: MFCD02683049
CAS-Nummer: 192130-34-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR