(5,6-Dichlor-3-pyridinyl)methanol 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: (5,6-Dichloropyridin-3-yl)methanol, 2,3-Dichloro-5-hydroxymethylpyridine, 5,6-Dichloronicotinyl Alcohol

OR17551-1G
APOSOR17551-1GEA 62.5 EUR
APOSOR17551-1G
(5,6-Dichlor-3-pyridinyl)methanol 95+%
(5,6-Dichlor-3-pyridinyl)methanol
Formel: C₆H₅Cl₂NO
Molecular Weight: 178,01 g/mol
Schmelzpunkt: 76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00671515
CAS-Nummer: 54127-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR