2-Chloro-6-methylpyrimidin-4yl-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-6-methylpyrimidin-4-ylamine
OR17556-1G
APOSOR17556-1GEA
53.8
EUR
APOSOR17556-1G
2-Chloro-6-methylpyrimidin-4yl-amine
2-Chloro-6-methylpyrimidin-4yl-amine
Formel:
C₅H₆ClN₃ Molecular Weight: 143,58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863311 CAS-Nummer: 14394-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...