1-(5-Nitropyridin-2-yl)piperazine 95%

Lieferant: Apollo Scientific
Achtung

OR1756-500MG
APOSOR1756-500MGEA 21.9 EUR
APOSOR1756-500MG
1-(5-Nitropyridin-2-yl)piperazine 95%
1-(5-Nitropyridin-2-yl)piperazine
Formel: C₉H₁₂N₄O₂
Molecular Weight: 208,2200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00053059
CAS-Nummer: 82205-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR