7-Bromchinolin

Lieferant: Apollo Scientific
Achtung

OR17561-25G OR17561-5G OR17561-1G
APOSOR17561-25GEA 87 EUR
APOSOR17561-25G APOSOR17561-5G APOSOR17561-1G
7-Bromchinolin
7-Bromoquinoline
Formel: C₉H₆BrN
Lagertemperatur: Raumtemperatur
MDL: MFCD03695823
CAS-Nummer: 4965-36-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR