7-Quinolinecarboxylic acid

Lieferant: Apollo Scientific
Achtung

OR17564-1G OR17564-250MG OR17564-5G
APOSOR17564-1GEA 95.7 EUR
APOSOR17564-1G APOSOR17564-250MG APOSOR17564-5G
7-Quinolinecarboxylic acid
7-Quinolinecarboxylic acid
Formel: C₁₀H₇NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD00047617
CAS-Nummer: 1078-30-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR