1-[(Benzyloxy)carbonyl]-2-piperidincarbonsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Benzyloxycarbonylpipecolic acid, 1-Carbobenzoxypipecolic acid, 1-Carbobenzoxy-2-piperidinecarboxylic acid, N-Cbz-2-Piperidinecarboxylic acid, Z-DL-Pip-OH, 1-Benzyloxycarbonyl-2-piperidinecarboxylic acid, 1-Cbz-pipecolic acid, 1-Cbz-2-piperidinecarboxylic acid

OR1757-100G
APOSOR1757-100GEA 122 EUR
APOSOR1757-100G
1-[(Benzyloxy)carbonyl]-2-piperidincarbonsäure 95%
N-Cbz-2-piperidinecarboxylic acid
Formel: C₁₄H₁₇NO₄
Molecular Weight: 263,2900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01863568
CAS-Nummer: 28697-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR