N-Boc-D-phenylalaninal 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-tert-Butyl (1-oxo-3-phenylpropan-2-yl)carbamate, (R)-(+)-2-(tert-Butoxycarbonylamino)-3-phenylpropanal, tert-butyl [(2R)-1-oxo-3-phenylpropan-2-yl]carbamate

OR17577-250MG
APOSOR17577-250MGEA 84.1 EUR
APOSOR17577-250MG
N-Boc-D-phenylalaninal 95%
N-Boc-D-phenylalaninal
Formel: C₁₄H₁₉NO₃
Molecular Weight: 249,31 g/mol
Schmelzpunkt: 86…89 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00274187
CAS-Nummer: 77119-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR