2-Nitro-m-xylol 99%

Lieferant: Apollo Scientific
Gefahr

OR17602-1KG OR17602-250G OR17602-4KG
APOSOR17602-1KGEA 0 EUR
APOSOR17602-1KG APOSOR17602-250G APOSOR17602-4KG
2-Nitro-m-xylol 99%
2-Nitro-m-xylol

Delivery of 1 or 4 kg quantity of this item to mainland UK is £35, additional charge will be added at order confirmation stage by our sales team.

Formel: C₈H₉NO₂
Siedepunkt: 225…227 °C
Schmelzpunkt: 14…16 °C
Dichte: 1,112
Flammpunkt: 88 °C
MDL: MFCD00007181
CAS-Nummer: 81-20-9
UN: 1665
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR