7-(Benzyloxy)-4-chloro-6-methoxyquinazoline 98%

Lieferant: Apollo Scientific
Achtung

OR17604-5G
APOSOR17604-5GEA 0 EUR
APOSOR17604-5G
7-(Benzyloxy)-4-chloro-6-methoxyquinazoline 98%
7-(Benzyloxy)-4-chloro-6-methoxyquinazoline
Formel: C₁₆H₁₃ClN₂O₂
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115120
CAS-Nummer: 162364-72-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR