1H-Indazole-5-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR17652-1G
APOSOR17652-1GEA 27.7 EUR
APOSOR17652-1G
1H-Indazole-5-carbaldehyde
1H-Indazole-5-carbaldehyde
Formel: C₈H₆N₂O
Molecular Weight: 146,1500 g/mol
Schmelzpunkt: 115…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06738280
CAS-Nummer: 253801-04-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR