2-(4-Methyl-2-phenylpiperazin-1-yl)nicotinic acid 97%

Lieferant: Apollo Scientific

Synonyme: Mirtazapine carboxylic acid

OR17666-250MG
APOSOR17666-250MGEA 53.8 EUR
APOSOR17666-250MG
2-(4-Methyl-2-phenylpiperazin-1-yl)nicotinic acid 97%
2-(4-Methyl-2-phenylpiperazin-1-yl)nicotinic acid
Formel: C₁₇H₁₉N₃O₂
Molecular Weight: 297,352 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458719
CAS-Nummer: 61338-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR