2-Bromo-1-(2-chloropyridin-3-yl)ethan-1-one 97%

Lieferant: Apollo Scientific

Synonyme: 3-(Bromoacetyl)-2-chloropyridine

OR17671-250MG
APOSOR17671-250MGEA 241 EUR
APOSOR17671-250MG
2-Bromo-1-(2-chloropyridin-3-yl)ethan-1-one 97%
2-Bromo-1-(2-chloropyridin-3-yl)ethan-1-one
Formel: C₇H₅BrClNO
Molecular Weight: 234,478 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13190086
CAS-Nummer: 237384-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR