6,6'-(9H-Fluorene-9,9-diyl)bis(2-naphthol) 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 9-Bis(6-hydroxy-2-naphthyl)fluorene, 6,6'-(9H-Fluorene-9,9-diyl)bis(naphthalen-2-ol)

OR17679-5G
APOSOR17679-5GEA 23.2 EUR
APOSOR17679-5G
6,6'-(9H-Fluorene-9,9-diyl)bis(2-naphthol) 95+%
6,6'-(9H-Fluorene-9,9-diyl)bis(2-naphthol)
Formel: C₃₃H₂₂O₂
Molecular Weight: 450,527 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD24038938
CAS-Nummer: 934557-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR