Methyl-(2E)-4-chlor-3-methoxy-2-butenoat 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(E)-4-Chloro-3-methoxy-2-butenoic Acid Methyl Ester, (E)-Methyl 4-chloro-3-methoxybut-2-enoate
OR17691-25G
APOSOR17691-25GEA
108
EUR
APOSOR17691-25G
Methyl-(2E)-4-chlor-3-methoxy-2-butenoat 95%
Methyl (E)-4-chloro-3-methoxy-2-butenoate
Formel:
C₆H₉ClO₃ Molecular Weight: 164,59 g/mol Schmelzpunkt: 30 °C Dichte: 1,21 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00071562 CAS-Nummer: 110104-60-4 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...