Ethyl-3-amino-1H-pyrazol-4-carboxylat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate

OR17692-100G
APOSOR17692-100GEA 133 EUR
APOSOR17692-100G
Ethyl-3-amino-1H-pyrazol-4-carboxylat 97%
Ethyl-3-amino-1H-pyrazol-4-carboxylat
Formel: C₆H₉N₃O₂
Molecular Weight: 155.16 g/mol
Schmelzpunkt: 103…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005238
CAS-Nummer: 6994-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR