(1H-Benzo[d]imidazol-2-yl)methanamine 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(Aminomethyl)-1H-benzimidazole

OR17701-100G OR17701-25G
APOSOR17701-100GEA 603 EUR
APOSOR17701-100G APOSOR17701-25G
(1H-Benzo[d]imidazol-2-yl)methanamine 95%
(1H-Benzo[d]imidazol-2-yl)methanamine
Formel: C₈H₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00464052
CAS-Nummer: 5805-57-2
UN: 3259
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR