2-Chloropyrimidine-4,5-diamine

Lieferant: Apollo Scientific
Achtung

OR17704-250MG
APOSOR17704-250MGEA 21.9 EUR
APOSOR17704-250MG
2-Chloropyrimidine-4,5-diamine
2-Chloropyrimidine-4,5-diamine
Formel: C₄H₅ClN₄
Molecular Weight: 144,5623 g/mol
Schmelzpunkt: 230…232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047355
CAS-Nummer: 14631-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR