2-Methyl-2-propanyl-N-(diphenylmethylen)glycinat 98%
Lieferant: Apollo Scientific
Synonyme:
N-(Diphenylmethylene)glycine tert-butyl ester, Glycine, N-(diphenylmethylene)-, 1,1-dimethylethyl ester, tert-Butyl 2-((diphenylmethylene)amino)acetate
OR17705-100G
APOSOR17705-100GEA
82.8
EUR
APOSOR17705-100G
2-Methyl-2-propanyl-N-(diphenylmethylen)glycinat 98%
tert-Butyl-2-((diphenylmethylene)amino)acetate
Formel:
C₁₉H₂₁NO₂ Molecular Weight: 295,382 g/mol Schmelzpunkt: 114…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134280 CAS-Nummer: 81477-94-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...