Quinoxaline-6-acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Quinoxalin-6-yl)acetic acid

OR17707-1G OR17707-250MG
APOSOR17707-1GEA 380 EUR
APOSOR17707-1G APOSOR17707-250MG
Quinoxaline-6-acetic acid
Quinoxaline-6-acetic acid
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188.19 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09880609
CAS-Nummer: 473932-16-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR