1H-Benzimidazol-5-boronsäurepinakolester 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzimidazole, Benzimidazole-5-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole

OR17711-1G
APOSOR17711-1GEA 66.7 EUR
APOSOR17711-1G
1H-Benzimidazol-5-boronsäurepinakolester 95+%
1H-Benzimidazol-5-boronsäurepinakolester
Formel: C₁₃H₁₇BN₂O₂
Molecular Weight: 244,10 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11054041
CAS-Nummer: 1007206-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR