1-Methyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester 98%

Lieferant: Apollo Scientific
Achtung

OR17713-1G OR17713-10G OR17713-5G
APOSOR17713-1GEA 21.9 EUR
APOSOR17713-1G APOSOR17713-10G APOSOR17713-5G
1-Methyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester 98%
1-Methyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester
Formel: C₁₂H₂₂BNO₂
Schmelzpunkt: 72…76 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11506069
CAS-Nummer: 454482-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR