(S)-(+)-2-Hydroxyisovaleriansäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutyric acid

OR17815-10G
APOSOR17815-10GEA 31.9 EUR
APOSOR17815-10G
(S)-(+)-2-Hydroxyisovaleriansäure 97%
(S)-(+)-2-Hydroxyisovaleriansäure
Formel: C₅H₁₀O₃
Molecular Weight: 118,13 g/mol
Siedepunkt: 124…125 °C/13mmHg
Schmelzpunkt: 68…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066443
CAS-Nummer: 17407-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR