3-Brom-5-methoxybenzonitril 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR17817-1G
APOSOR17817-1GEA
21.9
EUR
APOSOR17817-1G
3-Brom-5-methoxybenzonitril 95%
3-Brom-5-methoxybenzonitril
Formel:
C₈H₆BrNO Schmelzpunkt: 95…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08061971 CAS-Nummer: 867366-91-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...