Methyl-5-methyl-2-nitrobenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Nitro-m-toluic acid methyll ester, Methyl 6-Nitro-m-toluate, Methyl 5-methyl-2-nitrobenzoate, 5-Methyl-2-nitrobenzoic acid methyll ester
OR17822-1G
APOSOR17822-1GEA
21.9
EUR
APOSOR17822-1G
Methyl-5-methyl-2-nitrobenzoat
Methyl-5-methyl-2-nitrobenzoat
Formel:
C₉H₉NO₄ Molecular Weight: 195,1721 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00157275 CAS-Nummer: 20587-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...