4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-[4-(Chlorosulphonyl)phenyl]-3,5-dimethyl-1H-pyrazole

OR17844-1G
APOSOR17844-1GEA 296 EUR
APOSOR17844-1G
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzenesulphonyl chloride
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzenesulphonyl chloride
Formel: C₁₁H₁₁ClN₂O₂S
Molecular Weight: 270,735 g/mol
Schmelzpunkt: 75…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817557
CAS-Nummer: 61320-20-5
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR