6-Phenylpyridine-3-sulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR17846-1G
APOSOR17846-1GEA
377
EUR
APOSOR17846-1G
6-Phenylpyridine-3-sulphonyl chloride
6-Phenylpyridine-3-sulphonyl chloride
Formel:
C₁₁H₈ClNO₂S Schmelzpunkt: 106,5…107.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09064941 CAS-Nummer: 884507-12-4 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...