4-(1H-Imidazol-4-yl)aniline 95+%

Lieferant: Apollo Scientific

Synonyme: 4-(4-Aminophenyl)-1H-imidazole

OR17872-250MG
APOSOR17872-250MGEA 149 EUR
APOSOR17872-250MG
4-(1H-Imidazol-4-yl)aniline 95+%
4-(1H-Imidazol-4-yl)aniline
Formel: C₉H₉N₃
Molecular Weight: 159,188 g/mol
Schmelzpunkt: 97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00297065
CAS-Nummer: 29528-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR