2-Ethynylbenzoic acid 96+%

Lieferant: Apollo Scientific
Achtung

OR17875-5G
APOSOR17875-5GEA 230 EUR
APOSOR17875-5G
2-Ethynylbenzoic acid 96+%
2-Ethynylbenzoic acid
Formel: C₉H₆O₂
Molecular Weight: 146.143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203346
CAS-Nummer: 33578-00-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR