Methyl (2R,4R)-4-hydroxypyrrolidine-2-carboxylate hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R,4R)-Methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride, N-D-cis-Hyp-OMe·HCl, cis-4-Hydroxy-D-proline methyl ester hydrochloride, (2R,4R)-4-Hydroxy-2-(methoxycarbonyl)pyrrolidine hydrochloride

OR17883-1G
APOSOR17883-1GEA 21.9 EUR
APOSOR17883-1G
Methyl (2R,4R)-4-hydroxypyrrolidine-2-carboxylate hydrochloride
Methyl (2R,4R)-4-hydroxypyrrolidine-2-carboxylate hydrochloride
Formel: C₆H₁₁NO₃·ClH
Molecular Weight: 181.617 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00082536
CAS-Nummer: 114676-59-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR