5-Bromo-3-chloropyridine-2-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-3-chloropicolinonitrile

OR17892-1G
APOSOR17892-1GEA 21.9 EUR
APOSOR17892-1G
5-Bromo-3-chloropyridine-2-carbonitrile
5-Bromo-3-chloropyridine-2-carbonitrile
Formel: C₆H₂BrClN₂
Molecular Weight: 217,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12827550
CAS-Nummer: 945557-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR